Learn More
Electron Microscopy Sciences Flat Embedding Mold Flat Embed Mold Single End, 6 mm deep, 14 x 5 mm
SDP
Catalog No. 50930531
Encompass_Preferred
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. Standard flat embedding mold.
Depth: 6 mm
Size: 14 x 5 mm
Cavities: 21
Catalog No. 50-930-531 Supplier Electron Microscopy Sciences Supplier No. 70901
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. Standard flat embedding mold.
Depth: 6 mm
Size: 14 x 5 mm
Cavities: 21