Learn More
Electron Microscopy Sciences Flat Embedding Mold Flat Embed. Mold Triple Ends, 4 mm deep, 10 Cavities
SDP
Catalog No. 50930511
Encompass_Preferred
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. With 10 triple numbered, tapered tip cavities. Holds firmly in a chuck.
Depth: 4 mm
Cavities: 10
Catalog No. 50-930-511 Supplier Electron Microscopy Sciences Supplier No. 70875
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. With 10 triple numbered, tapered tip cavities. Holds firmly in a chuck.
Depth: 4 mm
Cavities: 10