Learn More
Electron Microscopy Sciences Flat Embedding Chuck
SDP
Catalog No. 5029557
Encompass_Preferred
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes.
Catalog No. 50-295-57 Supplier Electron Microscopy Sciences Supplier No. 69990
Precision machined from aluminum. These special chucks are made to accommodate small specimen blocks, which have been polymerized in Eppendorf tubes.