Learn More
Electron Microscopy Sciences Flat Embedding Mold Flat Embedding Mold Double Ends, 5 mm deep, 14 x 4 mm
SDP
Catalog No. 50930509
Encompass_Preferred
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. Flat mold has both ends tapered for embedding in both ends of each casting.
Depth: 5 mm
Size: 14 x 4 mm
Cavities: 21
Catalog No. 50-930-509 Supplier Electron Microscopy Sciences Supplier No. 70870
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. Flat mold has both ends tapered for embedding in both ends of each casting.
Depth: 5 mm
Size: 14 x 4 mm
Cavities: 21