Learn More
Electron Microscopy Sciences Flat Emery Stick 2/0 Grit
SDP
Catalog No. 50996757
Encompass_Preferred
Emery paper which is glued to a wooden handle. 11" long x 3/4" wide (279x19.1 mm). Very useful for filing excess resin of sample block tips.
Catalog No. 50-996-757 Supplier Electron Microscopy Sciences Supplier No. 6211902
Emery paper which is glued to a wooden handle. 11" long x 3/4" wide (279x19.1 mm). Very useful for filing excess resin of sample block tips.