Learn More
Grainger Flat-Free Wheel 4-1/2 350 lb.
Catalog No. 502767118
Encompass_Preferred
The Grainger 16" pneumatic flat free wheel has a load capacity of 1,500 lbs per wheel. It features a one-piece molded polyurethane tread that eliminates flats and provides excellent traction. The precision sealed ball bearings ensure smooth and quiet operation. This maintenance-free wheel fits 0.75" - 1.125" diameter stems and has a maximum speed of 5 mph. With dimensions of 16" x 4", this pneumatic flat free wheel is ideal for material handling applications where heavy loads, traction, and no maintenance are required.
Catalog No. 50-276-7118 Supplier Grainger Supplier No. 16V336
The Grainger 16" pneumatic flat free wheel has a load capacity of 1,500 lbs per wheel. It features a one-piece molded polyurethane tread that eliminates flats and provides excellent traction. The precision sealed ball bearings ensure smooth and quiet operation. This maintenance-free wheel fits 0.75" - 1.125" diameter stems and has a maximum speed of 5 mph. With dimensions of 16" x 4", this pneumatic flat free wheel is ideal for material handling applications where heavy loads, traction, and no maintenance are required.