Learn More
Thomas Scientific Vortexer Flat Head
SDP
Catalog No. 501461250
Encompass
Best Value! The BenchMixer™ sets the new standard in vortex mixing. With advanced Q-Drive™ technology, the BenchMixer provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. The unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Powerful motor for INSTANT vortexing Continuous or "touch" operation Q-Drive™ dynamic balancing system Variable speed from 200 to 3200 rpm Wide variety of accessories
Catalog No. 50-146-1250 Supplier Thomas Scientific Supplier No. 1227U59
Best Value! The BenchMixer™ sets the new standard in vortex mixing. With advanced Q-Drive™ technology, the BenchMixer provides smooth instant vortexing of even the largest sample sizes, including full 50ml tubes. The unique counter-balance system creates maximum vortexing action, while minimizing noise and excessive vibration. This results in quieter, more efficient and longer lasting operation. Other features include: "no-walk" suction feet, cold room/incubator compatibility and spill-proof electronic controls. Powerful motor for INSTANT vortexing Continuous or "touch" operation Q-Drive™ dynamic balancing system Variable speed from 200 to 3200 rpm Wide variety of accessories