Learn More
Electron Microscopy Sciences Lapping & Polishing Fixture Flat Mounting Blocks, 1.25", Stainless Steel
SDP
Catalog No. 50980970
Encompass
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with Models 150, 151, 155.
Diameter: 1.25"
Material: Stainless Steel
Catalog No. 50-980-970 Supplier Electron Microscopy Sciences Supplier No. 5015203
Add to Cart
Add to Cart
1" (25 mm) diameter stainless steel mounting blocks for lapping and polishing fixtures. For use with Models 150, 151, 155.
Diameter: 1.25"
Material: Stainless Steel