Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Flat PCR Caps, strips of 12

Catalog No. FERAB0850 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Natural
Product Type:
Flat Cap, 12-Cap Strip
PCR Plate Cap Strip
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
FERAB0850 Natural Flat Cap, 12-Cap Strip
AB-0851 Natural PCR Plate Cap Strip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERAB0850 Supplier Thermo Scientific™ Supplier No. AB0850
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.

Highlights

  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Certifications/Compliance Certified DNA/RNase Free
Material Polypropylene
Quantity 80 strips
Format 80 Strip Caps
For Use With (Equipment) Thermal Cycler
Product Type Flat Cap, 12-Cap Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.