Learn More
Grainger Flat Rack Nickel For 00680 Series
Catalog No. 502749392
Encompass_Preferred
The Grainger 14L719 is a 14-inch diameter, 719 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It also has a built-in backdraft damper to prevent reverse airflow. This powerful roof ventilator provides excellent ventilation for factories, warehouses, kitchens and other commercial spaces. With a max temperature rating of 104°F, it is suitable for most exhaust ventilation applications. The Grainger 14L719 roof exhaust fan delivers robust airflow for effective hot air removal.
Catalog No. 50-274-9392 Supplier Grainger Supplier No. 14L719
The Grainger 14L719 is a 14-inch diameter, 719 CFM, direct drive upblast roof exhaust fan. It features a 1/6 HP, 115V motor with permanently lubricated ball bearings and automatic thermal overload protection. The housing is constructed of heavy gauge aluminum with a spun aluminum venturi for maximum airflow. It also has a built-in backdraft damper to prevent reverse airflow. This powerful roof ventilator provides excellent ventilation for factories, warehouses, kitchens and other commercial spaces. With a max temperature rating of 104°F, it is suitable for most exhaust ventilation applications. The Grainger 14L719 roof exhaust fan delivers robust airflow for effective hot air removal.