Learn More
Electron Microscopy Sciences Flat Screw Cap Tubes Sterile, 15 ml, 17 x 118 mm
SDP
Catalog No. 501931048
Encompass_Preferred
The screw-cap conical tubes are sterile and repackaged into individual packets. The tubes are DNase, RNase, and pyrogen free with high contrast graduations, a white "frosted" marking area, and caps with a flat top surface for easy marking and labeling. Each tube is sterile right up until the moment you open the convenient peel pack.
Made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86°C. The maximum centrifugal force for the 15 ml tubes is 12,000 xg, while the 50 ml tubes can reach 15,000 xg.
Capacity: 15 ml
Size: 17 x 118 mm
Package: Individually Wrapped
Catalog No. 50-193-1048 Supplier Electron Microscopy Sciences Supplier No. 6302607
The screw-cap conical tubes are sterile and repackaged into individual packets. The tubes are DNase, RNase, and pyrogen free with high contrast graduations, a white "frosted" marking area, and caps with a flat top surface for easy marking and labeling. Each tube is sterile right up until the moment you open the convenient peel pack.
Made of premium polypropylene for clarity and centrifugal strength. Tubes and caps may be frozen at temperatures as low as -86°C. The maximum centrifugal force for the 15 ml tubes is 12,000 xg, while the 50 ml tubes can reach 15,000 xg.
Capacity: 15 ml
Size: 17 x 118 mm
Package: Individually Wrapped