Learn More
Sensorex Flat surface pH Sensor,DJ,BNC
Catalog No. 501897521
Encompass_Preferred
Flat measurement tip for surface measurements and slurries, double junction electrode design prevents reference contamination, durable Ultem body, booted BNC protects meter connection
Catalog No. 50-189-7521 Supplier Sensorex Supplier No. S350CD
Flat measurement tip for surface measurements and slurries, double junction electrode design prevents reference contamination, durable Ultem body, booted BNC protects meter connection