Learn More
MSC Vikan Hand Brush, Polyester Bristle, Medium, 6-1/4 in Head, Red
Catalog No. 502836938
Encompass
This hand brush is designed for scrubbing tasks, featuring durable polyester bristles and a plastic block. It has a medium bristle firmness and an overall length of 7 inches, making it suitable for various cleaning applications.
  • Polyester bristle material
  • Plastic block construction
  • Medium bristle firmness
  • 1 in bristle length
  • 7 in overall length
  • Red bristle color
  • 7 in head width
  • 6-1/4 in head length
  • Manufactured in Denmark
Catalog No. 50-283-6938 Supplier MSC Supplier No. 64425838
This hand brush is designed for scrubbing tasks, featuring durable polyester bristles and a plastic block. It has a medium bristle firmness and an overall length of 7 inches, making it suitable for various cleaning applications.
  • Polyester bristle material
  • Plastic block construction
  • Medium bristle firmness
  • 1 in bristle length
  • 7 in overall length
  • Red bristle color
  • 7 in head width
  • 6-1/4 in head length
  • Manufactured in Denmark