Learn More
Electron Microscopy Sciences Flat Bottom Embedding Capsules, Polyethylene 500/pk
SDP
Catalog No. 5025732
Encompass_Preferred
Added to our extensive line of embedding capsules is our flat bottom capsules, which are made from polyethylene, and are 8 mm (just like the standard 00 capsules) in diameter.
Catalog No. 50-257-32 Supplier Electron Microscopy Sciences Supplier No. 70021
Added to our extensive line of embedding capsules is our flat bottom capsules, which are made from polyethylene, and are 8 mm (just like the standard 00 capsules) in diameter.