Learn More
Analytical Sales & Services Flatpack Freezer Box for 12mm Tubes, 81 Wells - White
Catalog No. 5000806811
Encompass
Flatpack Freezer Box for 12mm Tubes, 81 Wells - White
Catalog No. 50-008-06811 Supplier Analytical Sales & Services Supplier No. FFB81W
Add to Cart
Add to Cart
Flatpack Freezer Box for 12mm Tubes, 81 Wells - White