Learn More
Analytical Sales & Services Flatpack Freezer Box for 12mm Tubes, 81 Wells
Catalog No. 5000805155
Encompass
Flatpack Freezer Box for 12mm Tubes, 81 Wells\n\n• Corrugated Polypropylene Box with Dividers\n• Store down to temperatures of -80°C\n\nQty/Pkg: 10
Catalog No. 50-008-05155 Supplier Analytical Sales & Services Supplier No. FFB81
Flatpack Freezer Box for 12mm Tubes, 81 Wells\n\n• Corrugated Polypropylene Box with Dividers\n• Store down to temperatures of -80°C\n\nQty/Pkg: 10