Learn More
Seca Corp 869 Digital flat scale with cable remote display
SDP
Supplier:  Seca Corp 8691321004
Encompass_Preferred
The robust seca 869 flat scale provides a 6.5 ft. long cable that connects base to the remote display. Additional convenience is supplied by the many extra weighing functions: BMI for nutritional conditions, HOLD to keep measurements displayed and TARE for weighing small children held in an adults arms. It is suitable for stationary or mobile use thanks to its handle, compact size and low weight. • Cable remote display for wall-mounting or table installation • Robust and durable • Support feet for strong stability • Can be powered with batteries (included) or optional power adapter seca model 400 • Capacity 550 lbs / 250 kg • Graduation 0.2 lb < 330 lbs > 0.5 lb / 100 g < 150 kg > 200 g • Functions: TARE, HOLD, BMI, lbs/kg switch-over, automatic switch-off
Catalog No. NC1599043