Learn More
Medchemexpress LLC Flavanomarein | 577-38-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN76755MG
Encompass_Preferred
Flavanomarein is a substance with cytoprotective, anti-inflammatory, and antioxidant activities. It enhances the phosphorylation level of AKT, regulates various protein expressions, and inhibits nuclear translocation of NF-κB p65. It also protects neuronal cells from damage and is applicable for studies related to Parkinson's disease and diabetic nephropathy.
  • Exhibits cytoprotective, anti-inflammatory, and antioxidant activities
  • Enhances the phosphorylation level of AKT
  • Regulates expression of PKC-δ, P85α, PKC-β1, Sirt1, Bcl-2, and ICAD
  • Inhibits the nuclear translocation of NF-κB p65
  • Regulates EMT marker proteins
  • Promotes the proliferation of HK-2 cells
  • Protects neuronal cells from 6-OHDA-induced neurotoxic damage
  • Applicable for studies related to Parkinson's disease and diabetic nephropathy
Catalog No. 50-004-18917