Learn More
Medchemexpress LLC HY-N7128 100mg Medchemexpress, Flavanone CAS:487-26-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7128100MG
Encompass_Preferred
Medchemexpress, HY-N7128 100mg Flavanone CAS:487-26-3 Flavanone is used to characterize the authenticity of lemon juice by monitoring of flavanone content. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0509