Learn More
Apexbio Technology LLC FLAVANONE-200MG
SDP
Catalog No. 5000570086
Encompass_Preferred
Flavanone (CAS No 487-26-3) is a key compound in the study of polyphenols primarily derived from citrus fruits and certain other plants It functions through various biological pathways notably by modulating antioxidant mechanisms influencing enzyme activity and interacting with cellular signaling pathways such as those involving kinases and transcription factors In vitro studies frequently demonstrate its activity in the nanomolar to low micromolar range showcasing its potential in modulating inflammatory responses and oxidative stress Flavanone is widely utilized in cell models to investigate these effects and is also employed in animal studies to explore its pharmacological potential including its efficacy in drug screening applications Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70086 Supplier Apexbio Technology LLC Supplier No. C6785200
Flavanone (CAS No 487-26-3) is a key compound in the study of polyphenols primarily derived from citrus fruits and certain other plants It functions through various biological pathways notably by modulating antioxidant mechanisms influencing enzyme activity and interacting with cellular signaling pathways such as those involving kinases and transcription factors In vitro studies frequently demonstrate its activity in the nanomolar to low micromolar range showcasing its potential in modulating inflammatory responses and oxidative stress Flavanone is widely utilized in cell models to investigate these effects and is also employed in animal studies to explore its pharmacological potential including its efficacy in drug screening applications Experimental concentrations typically depend on the specific research design