Learn More
Medchemexpress LLC Flavanone | 487-26-3 | 500 MG
SDP
Catalog No. 5000419772
Encompass_Preferred
Flavanone is a naturally occurring flavone with inhibitory activity for human estrogen synthetase (aromatase). It acts as an inhibitor for the ERK/p38/NF-κB signaling pathway and exhibits oral activity and antitumor efficacy.
  • Naturally occurring flavone.
  • Inhibits human estrogen synthetase (aromatase).
  • Inhibits the ERK/p38/NF-κB signaling pathway.
  • Exhibits oral activity.
  • Shows antitumor efficacy.
Catalog No. 50-004-19772 Supplier Medchemexpress LLC Supplier No. HYN7128500MG
Flavanone is a naturally occurring flavone with inhibitory activity for human estrogen synthetase (aromatase). It acts as an inhibitor for the ERK/p38/NF-κB signaling pathway and exhibits oral activity and antitumor efficacy.
  • Naturally occurring flavone.
  • Inhibits human estrogen synthetase (aromatase).
  • Inhibits the ERK/p38/NF-κB signaling pathway.
  • Exhibits oral activity.
  • Shows antitumor efficacy.