Learn More
Medchemexpress LLC HY-B1654A 25mg Medchemexpress, Flavin Adenine Dinucleotide Disodium CAS:84366-81-4 Purity:>98%
SDP
Catalog No. 501932983
Encompass_Preferred
Medchemexpress, HY-B1654A 25mg Flavin Adenine Dinucleotide Disodium CAS:84366-81-4 Flavin Adenine Dinucleotide Disodium is a redox cofactor, more specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2983 Supplier Medchemexpress LLC Supplier No. HYB1654A25MG
Medchemexpress, HY-B1654A 25mg Flavin Adenine Dinucleotide Disodium CAS:84366-81-4 Flavin Adenine Dinucleotide Disodium is a redox cofactor, more specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.