Learn More
Medchemexpress LLC Flavin adenine dinucleotide | 146-14-5 | 25 MG
SDP
Catalog No. 5000413912
Encompass_Preferred
Flavin adenine dinucleotide (Standard) is the analytical standard of Flavin adenine dinucleotide, intended for research and analytical applications. Flavin adenine dinucleotide (FAD) is a redox cofactor, specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for flavin adenine dinucleotide.
  • Intended for research and analytical applications.
  • Redox cofactor involved in important enzymatic reactions.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
Catalog No. 50-004-13912 Supplier Medchemexpress LLC Supplier No. 25MGHYB1654R
Flavin adenine dinucleotide (Standard) is the analytical standard of Flavin adenine dinucleotide, intended for research and analytical applications. Flavin adenine dinucleotide (FAD) is a redox cofactor, specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for flavin adenine dinucleotide.
  • Intended for research and analytical applications.
  • Redox cofactor involved in important enzymatic reactions.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).