Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flavin adenine dinucleotide | 146-14-5 | 98.9% | 100 UG
SDP

Catalog No. 5000413329
Encompass_Preferred

Flavin adenine dinucleotide (FAD) is a redox cofactor and a prosthetic group of a protein. It is involved in several crucial enzymatic reactions within metabolism.

  • Acts as a redox cofactor and a prosthetic group of a protein
  • Involved in several crucial enzymatic reactions within metabolism
  • Molecular weight: 785.55
  • Formula: C27H33N9O15P2
  • Appearance: Solid
  • Color: Yellow to orange
  • Initial source: Plants other families, also an endogenous metabolite, human gut microbiota metabolites, and found in microorganisms

Catalog No. 50-004-13329 Supplier Medchemexpress LLC Supplier No. HYB1654S100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flavin adenine dinucleotide (FAD) is a redox cofactor and a prosthetic group of a protein. It is involved in several crucial enzymatic reactions within metabolism.

  • Acts as a redox cofactor and a prosthetic group of a protein
  • Involved in several crucial enzymatic reactions within metabolism
  • Molecular weight: 785.55
  • Formula: C27H33N9O15P2
  • Appearance: Solid
  • Color: Yellow to orange
  • Initial source: Plants other families, also an endogenous metabolite, human gut microbiota metabolites, and found in microorganisms

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.