Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flavin adenine dinucleotide | 146-14-5 | MFCD00041835 | 785.55 g/mol | C27H33N9O15P2 | 10 MG
SDP

Catalog No. 5000297824
Encompass_Preferred

Flavin adenine dinucleotide (FAD) analytical standard supplied as a 10 mg reference material for research and analytical applications. FAD is a redox cofactor involved in multiple enzymatic reactions and is suitable as a reference for qualitative and quantitative HPLC, GC, and MS methods.

  • Analytical standard suitable for HPLC, GC, and MS assays.
  • Used as a quantitative reference for method development and validation.
  • CAS number 146-14-5; molecular formula C27H33N9O15P2.
  • Provided as a 10 mg vial; additional sizes available on request.
  • SDS and handling information available from the supplier.

Catalog No. 50-002-97824 Supplier Medchemexpress LLC Supplier No. HYB1654R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flavin adenine dinucleotide (FAD) analytical standard supplied as a 10 mg reference material for research and analytical applications. FAD is a redox cofactor involved in multiple enzymatic reactions and is suitable as a reference for qualitative and quantitative HPLC, GC, and MS methods.

  • Analytical standard suitable for HPLC, GC, and MS assays.
  • Used as a quantitative reference for method development and validation.
  • CAS number 146-14-5; molecular formula C27H33N9O15P2.
  • Provided as a 10 mg vial; additional sizes available on request.
  • SDS and handling information available from the supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.