Learn More
Medchemexpress LLC Flavin adenine dinucleotide (disodium salt) | 84366-81-4 | 99.7% | 50 MG
SDP
Catalog No. 5000413131
Encompass_Preferred
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, more specifically a prosthetic group of a protein. It is involved in several important enzymatic reactions in metabolism, making it crucial for various biological processes.
  • Functions as a redox cofactor
  • Acts as a prosthetic group for proteins
  • Participates in key enzymatic reactions in metabolism
  • Targeted as an endogenous metabolite
  • Relevant in metabolic enzyme and protease pathways
  • Applied in COVID-19 immunoregulation research
  • Used in cardiovascular disease research
Catalog No. 50-004-13131 Supplier Medchemexpress LLC Supplier No. HYB1654A50MG
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, more specifically a prosthetic group of a protein. It is involved in several important enzymatic reactions in metabolism, making it crucial for various biological processes.
  • Functions as a redox cofactor
  • Acts as a prosthetic group for proteins
  • Participates in key enzymatic reactions in metabolism
  • Targeted as an endogenous metabolite
  • Relevant in metabolic enzyme and protease pathways
  • Applied in COVID-19 immunoregulation research
  • Used in cardiovascular disease research