Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flavin adenine dinucleotide disodium salt | 84366-81-4 | 99.7% | 500 MG
SDP

Catalog No. 5000413091
Encompass_Preferred

Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor and a prosthetic group of a protein. It is involved in several important enzymatic reactions in metabolism.

  • Endogenous metabolite
  • Redox cofactor
  • Prosthetic group of a protein
  • Involved in several enzymatic reactions in metabolism
  • Appearance: solid
  • Color: yellow to orange
  • Molecular weight: 829.51
  • Formula: C27H31N9Na2O15P2

Catalog No. 50-004-13091 Supplier Medchemexpress LLC Supplier No. HYB1654A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor and a prosthetic group of a protein. It is involved in several important enzymatic reactions in metabolism.

  • Endogenous metabolite
  • Redox cofactor
  • Prosthetic group of a protein
  • Involved in several enzymatic reactions in metabolism
  • Appearance: solid
  • Color: yellow to orange
  • Molecular weight: 829.51
  • Formula: C27H31N9Na2O15P2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.