Learn More
Medchemexpress LLC Flavin adenine dinucleotide disodium salt | 84366-81-4 | MFCD00005041 | 99.72% | 500 UG
SDP
Catalog No. 5000413094
Encompass_Preferred
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, specifically a prosthetic group of a protein, that participates in several important enzymatic reactions in metabolism. It is for research use only.
  • Redox cofactor
  • Prosthetic group of a protein
  • Involved in several important enzymatic reactions in metabolism
Catalog No. 50-004-13094 Supplier Medchemexpress LLC Supplier No. HYB1654S500UG
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, specifically a prosthetic group of a protein, that participates in several important enzymatic reactions in metabolism. It is for research use only.
  • Redox cofactor
  • Prosthetic group of a protein
  • Involved in several important enzymatic reactions in metabolism