Learn More
Medchemexpress LLC Flavin adenine dinucleotide disodium salt | 84366-81-4 | MFCD00078174 | 99.7% | 1 ML
SDP
Catalog No. 5000413328
Encompass_Preferred
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism.
  • Endogenous metabolite
  • Redox cofactor
  • Involved in several important enzymatic reactions in metabolism
  • For research use only
Catalog No. 50-004-13328 Supplier Medchemexpress LLC Supplier No. HYB1654A10MM/1ML
Flavin adenine dinucleotide (FAD) disodium salt is a redox cofactor, specifically a prosthetic group of a protein, involved in several important enzymatic reactions in metabolism.
  • Endogenous metabolite
  • Redox cofactor
  • Involved in several important enzymatic reactions in metabolism
  • For research use only