Learn More
Medchemexpress LLC Flavin adenine dinucleotide disodium salt | 84366-81-4 | MFCD08277021 | >94.0% | 829.51 g/mol | C27H31N9Na2O15P2 | 10 MG
SDP
Catalog No. 5000298053
Encompass_Preferred
Flavin adenine dinucleotide (FAD) disodium salt is the disodium salt form of FAD, a redox cofactor and prosthetic group for flavoproteins involved in cellular metabolism. Supplied as a solid for research and analytical applications, it is commonly used in enzymology, redox assays, and biochemical studies.
  • Redox cofactor for flavoproteins and enzymatic reactions.
  • Suitable for research and analytical applications.
  • Available in small pack sizes for laboratory use.
  • Stable when stored sealed, away from moisture and light.
  • Well-characterized: CAS 84366-81-4; molecular weight 829.51 g/mol.
Catalog No. 50-002-98053 Supplier Medchemexpress LLC Supplier No. HYB1654A10MG
Flavin adenine dinucleotide (FAD) disodium salt is the disodium salt form of FAD, a redox cofactor and prosthetic group for flavoproteins involved in cellular metabolism. Supplied as a solid for research and analytical applications, it is commonly used in enzymology, redox assays, and biochemical studies.
  • Redox cofactor for flavoproteins and enzymatic reactions.
  • Suitable for research and analytical applications.
  • Available in small pack sizes for laboratory use.
  • Stable when stored sealed, away from moisture and light.
  • Well-characterized: CAS 84366-81-4; molecular weight 829.51 g/mol.