Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin Adenine Dinucleotide Disodium Salt, ≥95%, MP Biomedicals™

Catalog No. ICN15112783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15112783 250 mg
ICN15112790 500 mg
ICN15112791 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15112783 Supplier MP Biomedicals, Inc Supplier No. 0215112783
Add to Cart
Add to Cart
Cell Biology and Analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84366-81-4
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
PubChem CID 131675332
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Color Orange
Assay Percent Range ≥95%
Quantity 250 mg
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
Formula Weight 829.6
Physical Form Solid
Chemical Name or Material Flavin Adenine Dinucleotide Disodium Salt
EINECSNumber 205-663-1
Recommended Storage Store at 0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.