Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin Adenine Dinucleotide Disodium Salt Hydrate, ≥93% (dry basis)

Catalog No. AAJ1578503 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1578503 1 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ1578503 Supplier Thermo Scientific Supplier No. J1578503

missing translation for 'validMainframeMsg'

CAS: 1891059-93-0 | C27H31N9Na2O15P2 | 829.52 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1891059-93-0
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5 '-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
PubChem CID 131675332
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Physical Form Crystalline Powder
Quantity 1 g
IUPAC Name disodium;[[(2 R,3 S,4 R,5 R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2 R,3 S,4 S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
Formula Weight 829.51 (anhydrous)
Percent Purity ≥93%
Packaging Plastic bottle
Assay Dry basis
Chemical Name or Material Flavin Adenine Dinucleotide Disodium Salt Hydrate
EINECSNumber 282-733-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.