Learn More
Enzo Life Sciences Flavivirus group antigen Recombinant monoclonal antibody (D1-4G2-4-15 (4G2)) (Rabbit IgG kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4910200
Encompass_Preferred
Clone: D1-4G2-4-15 (4G2). Isotype: IgGκ. Immunogen: Dengue Virus type 2 antigens. Species reactivity: Virus. Specificity: Recognizes flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Yellow Fever Virus, Zika virus etc). Binds to the fusion loop at the extremity of domain II of protein E. Applications: ELISA, Flow Cytometry, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3017