Learn More
Medchemexpress LLC Flaviviruses-IN-3 | 420090-97-7 | C26H23N3O4S | 5 MG
SDP
Catalog No. 5000373011
Encompass_Preferred
Flaviviruses-IN-3 is a potent flavivirus inhibitor. It reduces West Nile virus (WNV) protease activity, showing an inhibition of 54%. This compound is intended for research use only.
  • Potent flavivirus inhibitor.
  • Reduces West Nile virus protease activity.
  • High purity: 98.93%.
  • Available in various quantities for research needs.
  • Provided with detailed documentation including data sheet, COA, SDS, and handling instructions.
  • Enables targeted research on flaviviruses.
  • Offers a tool for studying WNV protease activity.
  • Ensures reliable experimental results due to high purity.
  • Supports various experimental setups with different solubility protocols for in vitro and in vivo studies.
Catalog No. 50-003-73011 Supplier Medchemexpress LLC Supplier No. HY1530765MG
Flaviviruses-IN-3 is a potent flavivirus inhibitor. It reduces West Nile virus (WNV) protease activity, showing an inhibition of 54%. This compound is intended for research use only.
  • Potent flavivirus inhibitor.
  • Reduces West Nile virus protease activity.
  • High purity: 98.93%.
  • Available in various quantities for research needs.
  • Provided with detailed documentation including data sheet, COA, SDS, and handling instructions.
  • Enables targeted research on flaviviruses.
  • Offers a tool for studying WNV protease activity.
  • Ensures reliable experimental results due to high purity.
  • Supports various experimental setups with different solubility protocols for in vitro and in vivo studies.