Learn More
Cayman Chemical Flavokawain A, Purity: ≥98%, CAS: 37951-13-6, 10 mg
SDP
Catalog No. 502912820
Encompass_Preferred
A chalcone with diverse biological activities; activates Nrf2 and Hsf1 (EC50s = 14.1 and 7.9 µM, respectively, in reporter assays), increases GSH levels, and protects against hydrogen peroxide-induced cytotoxicity in HepG2 cells; inhibits LPS-induced increases in iNOS, COX-2, NO, and PGE2 levels in RAW 264.7 macrophages at 10 and 20 µM; induces apoptosis in T24 bladder cancer cells at 12.5 µg/ml; reduces tumor growth in an EJ bladder cancer mouse xenograft model at 50 mg/kg
Catalog No. 50-291-2820 Supplier Cayman Chemical Supplier No. 3608310 MG
A chalcone with diverse biological activities; activates Nrf2 and Hsf1 (EC50s = 14.1 and 7.9 µM, respectively, in reporter assays), increases GSH levels, and protects against hydrogen peroxide-induced cytotoxicity in HepG2 cells; inhibits LPS-induced increases in iNOS, COX-2, NO, and PGE2 levels in RAW 264.7 macrophages at 10 and 20 µM; induces apoptosis in T24 bladder cancer cells at 12.5 µg/ml; reduces tumor growth in an EJ bladder cancer mouse xenograft model at 50 mg/kg