Learn More
Medchemexpress LLC Flavokawain A | 3420-72-2 | 100 MG
SDP
Catalog No. 5000420271
Encompass_Preferred
Flavokawain A is a chalcone compound and an orally active inhibitor of PRMT5 and cytochrome P450. It exhibits anti-inflammatory, anti-tumor, and immunomodulatory effects. It can inhibit the proliferation of tumor cells and induce apoptosis and is used in research for diseases such as bladder cancer.
  • Inhibits proliferation of tumor cells and induces apoptosis
  • Exhibits anti-inflammatory, anti-tumor, and immunomodulatory effects
  • Used in research for diseases such as bladder cancer
  • Inhibitor of PRMT5 and cytochrome P450
Catalog No. 50-004-20271 Supplier Medchemexpress LLC Supplier No. HYN2420100MG
Flavokawain A is a chalcone compound and an orally active inhibitor of PRMT5 and cytochrome P450. It exhibits anti-inflammatory, anti-tumor, and immunomodulatory effects. It can inhibit the proliferation of tumor cells and induce apoptosis and is used in research for diseases such as bladder cancer.
  • Inhibits proliferation of tumor cells and induces apoptosis
  • Exhibits anti-inflammatory, anti-tumor, and immunomodulatory effects
  • Used in research for diseases such as bladder cancer
  • Inhibitor of PRMT5 and cytochrome P450