Learn More
Medchemexpress LLC Flavokawain A | 37951-13-6 | MFCD00017174 | 99.8% | 314.33 g/mol | C18H18O5 | 10 MG
SDP
Catalog No. 5000289691
Encompass_Preferred
(E)-Flavokawain A is a naturally occurring chalcone isolated from the kava plant, supplied as a research-grade standard for in vitro and preclinical studies. It exhibits reported anticarcinogenic activity through induction of apoptosis and is provided as a high-purity solid suitable for biochemical assays and cell biology research.
  • Chalcone compound with reported apoptosis-inducing activity
  • High purity: 99.8% (as supplied)
  • Empirical formula C18H18O5; molecular weight 314.33 g/mol
  • Available in small research quantities (e.g., 10 MG)
  • Store protected from light at 4°C; in solvent: -80°C for long-term storage
  • Intended for research use only; not for human or clinical use
Catalog No. 50-002-89691 Supplier Medchemexpress LLC Supplier No. HYN510610MG
(E)-Flavokawain A is a naturally occurring chalcone isolated from the kava plant, supplied as a research-grade standard for in vitro and preclinical studies. It exhibits reported anticarcinogenic activity through induction of apoptosis and is provided as a high-purity solid suitable for biochemical assays and cell biology research.
  • Chalcone compound with reported apoptosis-inducing activity
  • High purity: 99.8% (as supplied)
  • Empirical formula C18H18O5; molecular weight 314.33 g/mol
  • Available in small research quantities (e.g., 10 MG)
  • Store protected from light at 4°C; in solvent: -80°C for long-term storage
  • Intended for research use only; not for human or clinical use