Learn More
Lkt Laboratories Flavokawain B, ≥97%, 10MG
SDP
Supplier:  Lkt Laboratories F450310MG
Encompass_Preferred
Chalcone kavalactone that suppresses inflammation and inhibits degradation of IκBα and activation of NF-κB. CAS: 1775-97-9. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4919