Learn More
Cayman Chemical Flavokawain C, Purity: ≥98%, CAS: 37308-75-1, 10 mg
SDP
Catalog No. 502917182
Encompass_Preferred
A chalcone with enzyme inhibitory and anticancer activities; inhibits collagenase and elastase in cell-free assays (IC50s = 62.05 and 17.6 µM, respectively); cytotoxic to HCT116, HT-29, MCF-7, A549, and Ca Ski cancer cells (IC50s = 12.75, 39, 47.63, 40.28, and 39.88 µM, respectively); induces apoptosis and cell cycle arrest at the G1 phase in HCT116 cells at 20, 40, and 60 µM; reduces tumor volume and induces intratumoral necrosis and apoptosis in an HCT116 mouse xenograft model at 1 and 3 mg/kg
Catalog No. 50-291-7182 Supplier Cayman Chemical Supplier No. 3939310 MG
A chalcone with enzyme inhibitory and anticancer activities; inhibits collagenase and elastase in cell-free assays (IC50s = 62.05 and 17.6 µM, respectively); cytotoxic to HCT116, HT-29, MCF-7, A549, and Ca Ski cancer cells (IC50s = 12.75, 39, 47.63, 40.28, and 39.88 µM, respectively); induces apoptosis and cell cycle arrest at the G1 phase in HCT116 cells at 20, 40, and 60 µM; reduces tumor volume and induces intratumoral necrosis and apoptosis in an HCT116 mouse xenograft model at 1 and 3 mg/kg