Learn More
Medchemexpress LLC Flavokawain C | 37308-75-1 | MFCD00016489 | 99.5% | 300.31 g/mol | C17H16O5 | 10 MG
SDP
Catalog No. 5000289083
Encompass_Preferred
Flavokawain C is a naturally occurring chalcone isolated from Kava root that exhibits cytotoxic activity against human cancer cell lines (reported IC50 12.75 μM for HCT-116). It is supplied for research use and commonly used in in vitro studies of apoptosis, cell proliferation, and mechanism-of-action investigations.
  • Natural chalcone with reported apoptosis-inducing activity.
  • Reported IC50 12.75 μM against HCT-116 cells.
  • Molecular formula C17H16O5; molecular weight ~300.31 g/mol.
  • High reported purity (~99.5%).
  • Solid, light yellow to orange; store protected from light.
  • Available in small mg package sizes suitable for laboratory research.
Catalog No. 50-002-89083 Supplier Medchemexpress LLC Supplier No. HYN244510MG
Flavokawain C is a naturally occurring chalcone isolated from Kava root that exhibits cytotoxic activity against human cancer cell lines (reported IC50 12.75 μM for HCT-116). It is supplied for research use and commonly used in in vitro studies of apoptosis, cell proliferation, and mechanism-of-action investigations.
  • Natural chalcone with reported apoptosis-inducing activity.
  • Reported IC50 12.75 μM against HCT-116 cells.
  • Molecular formula C17H16O5; molecular weight ~300.31 g/mol.
  • High reported purity (~99.5%).
  • Solid, light yellow to orange; store protected from light.
  • Available in small mg package sizes suitable for laboratory research.