Learn More
Selleck Chemical LLC Flavone S3967-25mg
SDP
Catalog No. 503108688
Encompass_Preferred
Flavone (2-Phenylchromone 2-Phenyl-4-chromone 2-Phenyl-4-benzopyron) a class of flavonoids mainly found in spices and red or purple plant foods with antioxidant anti-proliferative anti-tumor anti-microbial estrogenic acetyl cholinesterase anti-inflammatory activities and are also used in cancer cardiovascular disease neurodegenerative disorders etc
Catalog No. 50-310-8688 Supplier Selleck Chemical LLC Supplier No. S396725MG
Flavone (2-Phenylchromone 2-Phenyl-4-chromone 2-Phenyl-4-benzopyron) a class of flavonoids mainly found in spices and red or purple plant foods with antioxidant anti-proliferative anti-tumor anti-microbial estrogenic acetyl cholinesterase anti-inflammatory activities and are also used in cancer cardiovascular disease neurodegenerative disorders etc