Learn More
Apexbio Technology LLC Flavone 525-82-6 25mg
SDP
Catalog No. 5000089203
Encompass_Preferred
Flavone (CAS 525-82-6) is a polyphenolic organic compound that serves as the structural prototype for the flavone subclass of flavonoids characterized by a 2-phenylchromen-4-one scaffold Flavone modulates diverse biological processes primarily through interactions with cellular signaling pathways and enzymes involved in oxidative stress and inflammation It is extensively utilized in biomedical research to investigate flavonoid metabolism antioxidant activity and anti-inflammatory mechanisms Flavone also functions as a reference standard in analytical assays and supports studies aiming to elucidate the pharmacological properties of related flavonoid derivatives
Catalog No. 50-000-89203 Supplier Apexbio Technology LLC Supplier No. C646425
Flavone (CAS 525-82-6) is a polyphenolic organic compound that serves as the structural prototype for the flavone subclass of flavonoids characterized by a 2-phenylchromen-4-one scaffold Flavone modulates diverse biological processes primarily through interactions with cellular signaling pathways and enzymes involved in oxidative stress and inflammation It is extensively utilized in biomedical research to investigate flavonoid metabolism antioxidant activity and anti-inflammatory mechanisms Flavone also functions as a reference standard in analytical assays and supports studies aiming to elucidate the pharmacological properties of related flavonoid derivatives