Learn More
Medchemexpress LLC HY-10005 10mg Medchemexpress, Flavopiridol CAS:146426-40-6 Purity:>98%
SDP
Catalog No. 502028605
Encompass_Preferred
Medchemexpress, HY-10005 10mg Flavopiridol CAS:146426-40-6 Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8605 Supplier Medchemexpress LLC Supplier No. HY1000510MG
Medchemexpress, HY-10005 10mg Flavopiridol CAS:146426-40-6 Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.