Learn More
Medchemexpress LLC Flavopiridol (alvocidib) | 146426-40-6 | 401.84 g/mol | C21H20ClNO5 | 500 MG
SDP
Catalog No. 5000215206
Encompass_Preferred
Flavopiridol is a broad-spectrum, ATP-competitive inhibitor of cyclin-dependent kinases used in research to probe cell-cycle regulation and kinase signaling. It inhibits CDK1, CDK2, and CDK4 with reported IC50s of 30 nM, 170 nM, and 100 nM, respectively. Supplied as a solid research reagent with documented solubility and storage recommendations.
Catalog No. 50-002-15206 Supplier Medchemexpress LLC Supplier No. HY10005500MG
Flavopiridol is a broad-spectrum, ATP-competitive inhibitor of cyclin-dependent kinases used in research to probe cell-cycle regulation and kinase signaling. It inhibits CDK1, CDK2, and CDK4 with reported IC50s of 30 nM, 170 nM, and 100 nM, respectively. Supplied as a solid research reagent with documented solubility and storage recommendations.