Learn More
Medchemexpress LLC Flavopiridol (Alvocidib) | 146426-40-6 | 99.4% | 401.84 | 100 MG
SDP
Catalog No. 5000364872
Encompass_Preferred
Flavopiridol (Alvocidib) is a broad-spectrum and competitive inhibitor of cyclin-dependent kinases (CDKs), effectively inhibiting CDK1, CDK2, and CDK4. It induces a distinct pattern of ER stress in CLL cells, contributing to cell death through IRE1-mediated activation of ASK1 and potentially downstream caspases. This compound also leads to potent upregulation of several primary response genes (PRGs) with both immediate and long-term effects on their expression. Furthermore, Flavopiridol inhibits cyclin E1 and induces apoptosis.
  • Broad-spectrum and competitive inhibitor of CDKs
  • Inhibits CDK1, CDK2, and CDK4
  • Induces ER stress in CLL cells
  • Contributes to cell death via IRE1-mediated activation of ASK1
  • Upregulates primary response genes
  • Inhibits cyclin E1
  • Induces apoptosis
Catalog No. 50-003-64872 Supplier Medchemexpress LLC Supplier No. 100MGHY10005
Flavopiridol (Alvocidib) is a broad-spectrum and competitive inhibitor of cyclin-dependent kinases (CDKs), effectively inhibiting CDK1, CDK2, and CDK4. It induces a distinct pattern of ER stress in CLL cells, contributing to cell death through IRE1-mediated activation of ASK1 and potentially downstream caspases. This compound also leads to potent upregulation of several primary response genes (PRGs) with both immediate and long-term effects on their expression. Furthermore, Flavopiridol inhibits cyclin E1 and induces apoptosis.
  • Broad-spectrum and competitive inhibitor of CDKs
  • Inhibits CDK1, CDK2, and CDK4
  • Induces ER stress in CLL cells
  • Contributes to cell death via IRE1-mediated activation of ASK1
  • Upregulates primary response genes
  • Inhibits cyclin E1
  • Induces apoptosis