Learn More
Medchemexpress LLC HY-10006 5mg Medchemexpress, Flavopiridol (Hydrochloride) CAS:131740-09-5 Purity:>98%
SDP
Catalog No. 501931709
Encompass_Preferred
Medchemexpress, HY-10006 5mg Flavopiridol (Hydrochloride) CAS:131740-09-5 Flavopiridol Hydrochloride is a broad inhibitor of CDK , competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC 50 s of 30, 170, 100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1709 Supplier Medchemexpress LLC Supplier No. HY100065MG
Medchemexpress, HY-10006 5mg Flavopiridol (Hydrochloride) CAS:131740-09-5 Flavopiridol Hydrochloride is a broad inhibitor of CDK , competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC 50 s of 30, 170, 100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.