Learn More
Medchemexpress LLC HY-10006 100mg , Flavopiridol (Hydrochloride) HL 275;NSC 649890;MDL 107826A;FLAVOPIRIDOL HCL;Alvocidib Hydrochloride CAS:131740-09-5 Purity:98%
SDP
Catalog No. 501492860
Encompass_Preferred
Medchemexpress, HY-10006 100mg Flavopiridol (Hydrochloride) HL 275;NSC 649890;MDL 107826A;FLAVOPIRIDOL HCL;Alvocidib Hydrochloride CAS:131740-09-5 Formula:C21H21Cl2NO5 IC50: 30 nM (CDK1), 170 nM (CDK2), 100 nM (CDK4) Purity:98% Flavopiridol Hydrochloride is a broad inhibitor of CDK , competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC 50 s of 30, 170, 100 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2860 Supplier Medchemexpress LLC Supplier No. HY10006100MG
Medchemexpress, HY-10006 100mg Flavopiridol (Hydrochloride) HL 275;NSC 649890;MDL 107826A;FLAVOPIRIDOL HCL;Alvocidib Hydrochloride CAS:131740-09-5 Formula:C21H21Cl2NO5 IC50: 30 nM (CDK1), 170 nM (CDK2), 100 nM (CDK4) Purity:98% Flavopiridol Hydrochloride is a broad inhibitor of CDK , competing with ATP to inhibit CDKs including CDK1, CDK2, CDK4 with IC 50 s of 30, 170, 100 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.