Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flavopiridol hydrochloride (alvocidib hydrochloride) | 131740-09-5 | MFCD26792554 | 438.30 g/mol | C21H21Cl2NO5 | 10 MG
SDP

Catalog No. 5000215187
Encompass_Preferred

Flavopiridol hydrochloride (alvocidib) is an ATP-competitive, broad-spectrum cyclin-dependent kinase (CDK) inhibitor used in research to probe cell-cycle control, autophagy, and viral pathways. The hydrochloride salt enhances solubility for in vitro and in vivo applications; manufacturer datasheet provides IC50s, solubility, storage, and formulation guidance for stock and dosing preparation.

  • ATP-competitive cyclin-dependent kinase inhibitor.
  • Reported IC50s: CDK1 30 nM, CDK2 170 nM, CDK4 100 nM.
  • High solubility in DMSO and water (≥20 mg/mL) for stock solutions.
  • Includes in vivo formulation guidance for dosing preparations.
  • Stability in solvent: -80°C (1 year) and -20°C (6 months) when sealed.
  • Molecular weight 438.30 g/mol and formula C21H21Cl2NO5 for accurate calculations.

Catalog No. 50-002-15187 Supplier Medchemexpress LLC Supplier No. HY1000610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flavopiridol hydrochloride (alvocidib) is an ATP-competitive, broad-spectrum cyclin-dependent kinase (CDK) inhibitor used in research to probe cell-cycle control, autophagy, and viral pathways. The hydrochloride salt enhances solubility for in vitro and in vivo applications; manufacturer datasheet provides IC50s, solubility, storage, and formulation guidance for stock and dosing preparation.

  • ATP-competitive cyclin-dependent kinase inhibitor.
  • Reported IC50s: CDK1 30 nM, CDK2 170 nM, CDK4 100 nM.
  • High solubility in DMSO and water (≥20 mg/mL) for stock solutions.
  • Includes in vivo formulation guidance for dosing preparations.
  • Stability in solvent: -80°C (1 year) and -20°C (6 months) when sealed.
  • Molecular weight 438.30 g/mol and formula C21H21Cl2NO5 for accurate calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.