Learn More
Apexbio Technology LLC Flavoxate hydrochloride 3717-88-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094220
Encompass_Preferred
Flavoxate hydrochloride (CAS 3717-88-2) is a synthetic small molecule that functions as an antagonist of muscarinic acetylcholine receptors (AChRs) By inhibiting muscarinic receptor-mediated signaling it disrupts smooth muscle contractions within the urinary tract Flavoxate hydrochloride is widely utilized in biomedical research to investigate mechanisms underlying bladder overactivity urinary tract disorders and cholinergic signaling pathways Its antispasmodic properties support studies focused on smooth muscle pharmacology and the modulation of acetylcholine-mediated responses
Catalog No. 50-000-94220 Supplier Apexbio Technology LLC Supplier No. B34335.1
Flavoxate hydrochloride (CAS 3717-88-2) is a synthetic small molecule that functions as an antagonist of muscarinic acetylcholine receptors (AChRs) By inhibiting muscarinic receptor-mediated signaling it disrupts smooth muscle contractions within the urinary tract Flavoxate hydrochloride is widely utilized in biomedical research to investigate mechanisms underlying bladder overactivity urinary tract disorders and cholinergic signaling pathways Its antispasmodic properties support studies focused on smooth muscle pharmacology and the modulation of acetylcholine-mediated responses