Learn More
Medchemexpress LLC Flavoxate (hydrochloride) | 3717-88-2 | 99.7% | 25 MG
SDP
Catalog No. 5000450820
Encompass_Preferred
Flavoxate hydrochloride (Standard) is an analytical reference standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. It is an orally active L-type Ca2+ channel inhibitor and antispasmodic agent. It functions as a potent and competitive phosphodiesterase inhibitor and a muscarinic mAChR antagonist, demonstrating moderate calcium antagonistic activity and local anesthetic effect.
  • Inhibits cyclic adenosine monophosphate production
  • Regulates the brainstem micturition center
  • Stimulates G protein-coupled receptors
  • Relaxes bladder smooth muscle
  • Inhibits isovolumetric rhythmic contractions
  • Increases bladder capacity
  • Reduces urgent urination frequency and pollakiuria symptoms
  • Suitable for overactive bladder and lower urinary tract infection research
Catalog No. 50-004-50820 Supplier Medchemexpress LLC Supplier No. 25MGHYB0549AR
Flavoxate hydrochloride (Standard) is an analytical reference standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. It is an orally active L-type Ca2+ channel inhibitor and antispasmodic agent. It functions as a potent and competitive phosphodiesterase inhibitor and a muscarinic mAChR antagonist, demonstrating moderate calcium antagonistic activity and local anesthetic effect.
  • Inhibits cyclic adenosine monophosphate production
  • Regulates the brainstem micturition center
  • Stimulates G protein-coupled receptors
  • Relaxes bladder smooth muscle
  • Inhibits isovolumetric rhythmic contractions
  • Increases bladder capacity
  • Reduces urgent urination frequency and pollakiuria symptoms
  • Suitable for overactive bladder and lower urinary tract infection research